CellPro™ Disposable Cell Spreaders
Supplier: Alkali Scientific
TS5420 TS5421 TS5430 TS5431 TS5440 TS5441 TS5450 TS5451
104028-028EA 135.37 USD
104028-028 104026-636 104027-866 104028-346 104028-334 104029-038 104027-492 104027-938
CellPro™ Disposable Cell Spreaders
Spreaders Cell Spreaders
Facilitate cell culture procedures with Alkali Scientific™ disposable cell spreaders, designed for easy and consistent cell spreading in laboratory applications.
  • Polypropylene
  • 4 different shapes and sizes
  • Original L-shape, a T-shape version and two triangle shapes (large and small)
  • All supplied in sterile packaging
  • Autoclavable and have smooth, rounded edges to prevent unwanted cutting, scraping or gouging of agar during inoculation
The L-shape spreaders are ideal for spreading bacteriological cultures evenly. The shape allows the user to rotate and apply easily throughout an entire round plate or dish. They are made of ABS and have a slight upward curve at the edge to prevent dragging or scraping. The T-shape spreaders ensure that consistently even pressure can be applied across the entire span of the 34 mm spreading surface. Designed for spreading and dispersing liquids across the surface of agar culture plates, they are made of ABS and commonly used for CFU plate counts on water, milk, urine, etc. Triangle shape spreaders are also used for CFU count applications, but have a grooved handle that improves grip and orientation. Made of polypropylene, they are available with a 30 mm span or an extra large 60 mm span.