CellPro™ Premium Flat Bottom Cell Culture Plates with Lids, TC Treated, Sterile
Supplier: Alkali Scientific
TPN1006 TPN1012 TPN1024 TPN1048 TPN1096
104028-376EA 110.75 USD
104028-376 104027-752 104028-656 104028-494 104028-734
CellPro™ Premium Flat Bottom Cell Culture Plates with Lids, TC Treated, Sterile
Microplates Cell Culture Plates
Culture cells with optimal adherence in the Alkali Scientific™ CellPro™ premium flat bottom cell culture plates, featuring TC treatment for enhanced cell attachment, supplied sterile with lids for convenient use.
  • High clarity, 100% virgin polystyrene
  • Vacuum gas plasma tissue culture treatment
  • Clear lot number for batch tractability
  • Peel-to-open, individually wrapped and sterilized by ga mma irradiation
  • Markings of well coordinates available for 12, 24, 48 and 96 well plates
  • Non-pyrogenic, DNase/RNase free
  • Gamma sterilized
Elevate your cell culture experiments with CellPro™ premium flat bottom cell culture plates from Alkali Scientific™. These plates, meticulously designed for optimal cell growth and maintenance, feature a flat bottom and are treated with TC for superior cell adhesion and proliferation. Each plate comes with a secure lid, ensuring sample integrity and maintaining sterility throughout your experiments. With their premium quality and sterile packaging, these cell culture plates provide a reliable platform for a wide range of cell-based assays and studies.