Cell Culture Flasks, Vent Cap, Non-Treated, Sterile
Supplier: Alkali Scientific
TL31 TL32 TL33 TL34
104026-676EA 172.29 USD
104026-676 104029-110 104028-014 104027-628
Cell Culture Flasks, Vent Cap, Non-Treated, Sterile
Flasks Cell Culture Flasks
Cultivate cells effectively with Alkali Scientific™ Cell Culture Flasks, featuring a vent cap design and sterile packaging for convenient cell culture applications.
  • Packaged in sterile, resealable zip bags
  • High clarity, 100% virgin polystyrene
  • Frosted writing and clear graduations
  • Stackable
  • Sterilized by E-Beam
  • Vent caps (with breathable 0.22 um membrane)
  • Clear lot number for batch traceability
  • Non-pyrogenic, DNase/RNase free
Alkali Scientific™ presents cell culture flasks designed for optimal cell growth and maintenance. These flasks feature vent caps to facilitate gas exchange, ensuring optimal conditions for cell culture. Manufactured under sterile conditions, they are ready for immediate use in your laboratory. With a non-treated surface, these flasks provide versatility for various cell culture applications, allowing researchers to tailor culture conditions according to their specific needs. Whether you're propagating cell lines or conducting experiments, trust Alkali Scientific™ cell culture flasks to deliver consistent and reliable results.