Cell Culture Lab Dishes
Supplier: Alkali Scientific
TD0035 TD0060 TD0100 TD0125 TD0150
104027-706EA 161.46 USD
104027-706 104027-028 104027-270 104027-876 104028-556
Cell Culture Lab Dishes
Culture Dishes Cell Culture Dishes
Perform a variety of cell culture tasks with Alkali Scientific™ Cell Culture Lab Dish, ideal for routine laboratory procedures.
  • Optically clear for microscopy applications
  • Large pronounced gripping rings
  • Air exchange vents
  • Pyrogen-free
  • Gamma sterilized
  • Tissue culture treated
  • Unique lid inner design for effective gas e×change
  • External grip for better handling
  • Excellent stackability
  • USP guideline compliant (USP class VI tested)
  • Non - pyrogenic, non - cytotoxic, DNase/RNase-free, and human DNA-free
The Alkali Scientific™ Cell Culture Lab Dish represents an essential component for cell culture experimentation within laboratory environments. Constructed from premium-quality plastic materials, these dishes feature a flat bottom and a meticulously smooth surface, facilitating optimal cell adhesion and proliferation. With their generous surface area, they acco mmodate large cell populations, making them ideal for diverse applications ranging from routine cell maintenance to advanced cellular assays. Designed to sea mlessly integrate with standard laboratory equipment, these dishes offer versatility and reliability, empowering researchers to conduct experiments with precision and confidence.