StripFilm™ 1×8-Polypropylene Strip Sealing Film
Supplier: Alkali Scientific
SF8 SF8-S
104028-596EA 44.6 USD
104028-596 104027-154
StripFilm™ 1×8-Polypropylene Strip Sealing Film
Seals Microplate Seals
Ensure tight seals for PCR plates with 1×8-StripFilm™ Polypropylene Strip Sealing Film, provided in a pack of 400 for convenience.
  • Certified DNase- and RNase- free
  • Designed to be used with both polystyrene and polypropylene plates
  • Precision cut to a smaller dimension to seal a single 8-well row of a 96-well plate or an ELISA strip-well plate
  • Each sheet contains 8 Mini Strips on a continuous backing liner for ease of use
  • Reco mmended for temperature range –40 to +120 °C
  • Mini Strips can be peeled and applied individually or two-at-a-time for increased efficiency
  • Mini Strips can be used to selectively cover one row of a plate to prevent accidental pipetting or to re-seal rows that have been pierced
  • 2.0 mil polypropylene film features good clarity and easy handling
  • Specially formulated adhesive provides an e×cellent seal to minimize evaporation, reduce cross-contamination and prevent spillage
  • Removes easily and cleanly from plate surfaces
  • Minimizes aerosols upon film removal
  • Features two end-tabs with perforations for ease of handling, application and removal
For sealing only one 8-well row of a 96-well plate. 8-Strips per sheet, sterile and non-sterile.
Designed for use with either 1×8-StripFilm™ ELISA plates or standard 96-well plates where rows must be selectively protected or accessed. Ideal for COVID-19 testing to seal a row of 8 wells after swabs are deposited into a deep-well plate.
Packaging: 400 strips.