96 Well Sealing Mats
Supplier: Alkali Scientific
MPD54-S MPD51 MPD51-S MPD52 MPD52-S MPD53-S
104027-404EA 247.68 USD
104027-404 104027-772 104027-546 104027-844 104027-710 104027-558
96 Well Sealing Mats
Seals Microplate Seals
Keep samples securely sealed with Alkali Scientific™ 96 well sealing mats, available in packs of 50.
  • Alphanumeric grid-referencing
  • Molded from a proprietary thermoplastic elastomer for a flexible design
  • Suitable for long term storage
  • Available in autoclavable or pre-slit pierceable formats
  • Solvent resistant (including DMSO, ethanol, and isopropanol)
  • Suitable for UV irradiation and ga mma irradiation
  • Requires 6N for piercing
  • Temperature range: −20 to +55 °C
Ensure the integrity of your samples with the Alkali Scientific™ 96 well sealing mats. Designed for use with 96-well plates, these sealing mats provide a secure seal to prevent evaporation and contamination during storage or analysis. Each case contains 50 sealing mats, offering ample supply for your laboratory needs. Trust in the reliability and performance of these sealing mats to safeguard your valuable samples and maintain the accuracy of your experimental results.