Inoculating Loops and Needles
Supplier: Alkali Scientific
M2015 M2025 M2030
104027-182EA 104.6 USD
104027-182 104028-030 104027-730
Inoculating Loops and Needles
Spreaders Inoculating Loops and Needles
Perform accurate streaking and inoculation with Alkali Scientific inoculating loops and needles, available in a pack of 1000 for microbiological applications.
  • Sterile
  • Uniform streaking and inoculating
  • Hexagonal grip
  • Smooth loop surfaces - no gouging
  • Non toxic, biologically inert polypropylene
The inoculating loops and needles set offers a reliable and versatile solution for microbiological inoculation and streaking procedures in laboratory settings. With 1000 pieces per pack, this comprehensive set ensures ample supply for routine microbiology work and experimentation. Crafted from high-quality materials, including durable metals and heat-resistant plastics, the loops and needles are designed to withstand repeated sterilization cycles, ensuring optimal performance and longevity. The precision-engineered design of each loop and needle facilitates accurate and consistent transfer of microbial cultures onto agar plates or culture media, minimizing the risk of contamination and ensuring reliable results. Whether used in clinical diagnostics, research laboratories, or educational institutions, the inoculating loops and needles set provides microbiologists and laboratory technicians with the tools they need for efficient and precise microbiological work.