CellPro™ Tube Storage Box, Hinged lid
Supplier: Alkali Scientific
FS100-BK FS100-C FS100-G FS100-R FS100-Y FS100-A FS100-B
104027-998EA 55.37 USD
104027-998 104028-870 104027-662 104028-026 104028-878 104027-870 104027-538
CellPro™ Tube Storage Box, Hinged lid
Boxes Cooling/Freezing Boxes and Bags
Store tubes securely with Alkali Scientific Tube Storage Box, featuring a hinged lid for easy access and storage convenience.
  • Polypropylene, withstands –80 ℃
  • 6 types - for 0.2 to 50 ml tubes
  • Compact and space efficient
  • Autoclavable
  • Numbered tube wells
Streamline your sample storage with Alkali Scientific Tube Storage Box featuring a hinged lid design. This user-friendly storage solution ensures secure and organized storage for your tubes, while the hinged lid eliminates the hassle of misplacing or losing separate lids. The robust construction andMore Product Information