Stainless Steel Autoclave and Incubator Rack
Supplier: Alkali Scientific
AR3015
104027-268EA 132.29 USD
104027-268
Stainless Steel Autoclave and Incubator Rack
Racks Incubator Racks
The Alkali Scientific™ stainless steel autoclave and incubator rack is engineered to meet the stringent requirements of modern laboratory environments. Constructed from high-grade stainless steel, this rack offers exceptional resistance to corrosion and degradation under high-temperature and high-humidity conditions, making it suitable for autoclave sterilization and incubation applications.
  • Stainless steel
  • Holds 41×15 ml tubes
The rack's design ensures optimal stability and support for various laboratory instruments and samples. Its robust construction enhances durability, maintaining structural integrity through repeated cycles of sterilization and use. This rack is an essential tool for maintaining organized and efficient laboratory workflows, ensuring consistent and reliable performance across diverse experimental conditions.
Alkali Scientific's dedication to precision engineering and quality assurance is evident in the meticulous design and fabrication of this stainless-steel rack, making it a reliable component for any laboratory setting.