LabKlenz™ 110 Alkaline Detergents
Supplier: Steris
LabKlenz™
1L1108 1L1102 1L1105 1L1108
89089-202CS 288.7 USD
CA89089-202 89089-202 89089-200 89089-198
LabKlenz™ 110 Alkaline Detergents
Cleaning Agents Detergents and Surface Cleansers
LabKlenz™ 110 Detergent is a phosphate-free, liquid, alkaline detergent for use in automated laboratory glassware washing applications. It is extremely free rinsing to meet critical laboratory requirements.
  • Flexibility - Highly alkaline for effective removal of residues in both soft and hard water
  • Cost Savings - Reduces utility consumption and cleaning time due with low foaming, free-rinsing formulation
  • Environmentally Friendly - phosphate-free formulation with biodegradable surfactants
  • Reliability - Manufactured in a cGMP compliant, FDA registered and ISO 9001 certified facility
LabKlenz™ 110 Detergent is specially formulated to meet the unique cleaning and materials compatibility requirements found in laboratory glassware washing applications.