Sabouraud Dextrose Broth

Supplier: RPI

S12520-1000.0 S12520-500.0
77510-418EA 236.48 USD
77510-418 77510-420
Sabouraud Dextrose Broth
Media for Microbiology Dehydrated Media for Microbiology

Dissolve 30 g in 1 L distilled water. Heat and stir to completely dissolve agar. Sterilize by autoclaving for 15 minutes.


Formulation:
Casein Digest Peptone: 10.0 g/L
Dextrose: 20.0 g/L


Caution: Research or further manufacturing use only, not for food or drug use.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR