Lactobacillus MRS Broth Powder
Supplier: RPI
L11000-1000.0 L11000-500.0 L11000-2500.0
76346-420EA 212.05 USD
76346-420 76346-422 77510-294
Lactobacillus MRS Broth Powder
Media for Microbiology Dehydrated Media for Microbiology
Recommended for the selective isolation and enumeration of Lactobacilli.
Caution: Research or further manufacturing use only, not for food or drug use.