Bottled Media, Teknova
Supplier: TEKNOVA INC
T7655 T7655 C0860 C0860 N0165 C4014 L5150 F0480-06 L8950 N1215 C4014 S1635
100220-294EA 72.74 USD
100220-294 101319-698 100217-486 100220-270 200066-600 CA100220-270 100220-306 100220-244 CA101414-090 101414-090 100216-502 CA101319-698
Bottled Media, Teknova
For use as a carbon source in growth media.
  • 50% Glucose Solution
  • Sterile
Ship and store at room temperature.