D-(+)-Sucrose ACS

Supplier: RPI

S24065-1000.0 S24065-2500.0 S24065-3000.0 S24065-500.0 S24065-5000.0 S24065-10000.0
76348-234EA 56.67 USD
76348-234 76348-236 76348-238 76348-240 76348-242 77510-438
D-(+)-Sucrose ACS
D(+)-Sucrose
Formula: C₁₂H₂₂O₁₁
Storage Temperature: Room Temperature
MDL Number: MFCD00006626
CAS Number: 57-50-1
EINECS: 200-334-9

Order Now

Specification Test Results

Appearance Fine White Crystals
Specific Rotation +66.3 - +66.8 °
Insoluble ≤ 0.005 %
Loss on Drying (105 oC) ≤ 0.03 %
Residue after Ignition ≤ 0.01 %
Titrable Acid ≤ 0.0008 meq/g
Chloride (Cl) ≤ 0.005 %
Sulfate and Sulfite (SO4) ≤ 0.005 %
Heavy Metals (as Pb) ≤ 0.0005 %
Iron ≤ 0.0005 %
Invert Sugar ≤ 0.05 %
Source Sugar beets or sugar cane
BSE/TSE Free Conforms


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR