Acylation Reagent, MBTFA
Supplier: Thermo Scientific
TS-49700
10066-374EA 449.68 USD
10066-374
Acylation Reagent, MBTFA
Chromatography Sorbents GC Sorbents
Trifluoroacylate primary and secondary amines, hydroxyl and thiol groups, and carbohydrates with the Thermo Scientific™ MBTFA Acylation Reagent. The principal byproduct N-methyltrifluoroacetamine is stable, volatile, and does not present problems in subsequent GC. MBTFA produces very volatile derivatives of carbohydrates and can be used to selectively acylate amines in the presence of hydroxyl and carboxyl groups that have been protected by silylation.
  • Reacts under nonacidic conditions
  • Principle byproduct from the derivatization reaction is N-methyltrifluoroacetamine, which is stable, volatile and does not present problems in subsequent GC
  • Produces very volatile derivatives of carbohydrates
  • Can be used to selectively acylate amines in the presence of hydroxyl and carboxyl groups that have been protected by silylation
Caution: For research use only. Not for use in diagnostic procedures.