UltraSOFT 5000™ Powder-Free Nitrile Gloves with Textured Fingertips
Supplier: Alkali Scientific
NG2205 NG2210 NG2215 NG2220 NG2225
104028-188EA 410.75 USD
104028-188 104028-882 104027-764 104027-994 104027-002
UltraSOFT 5000™ Powder-Free Nitrile Gloves with Textured Fingertips
Gloves
UltraSOFT 5000™ Powder-free nitrile exam gloves have a soft, comfortable feel and feature textured fingertips for a strong grip. These latex-free nitrile gloves are great for people who have latex allergies, and they are ideal for use in laboratory or clinical settings.
  • Highly elastic comfortable feel
  • Powder-free with textured fingertips
  • 300 gloves per box; 3 times more per case
  • Non-sterile
  • Medical Examination Grade, FDA 510(k)
  • Tensile strength on spec sheet: 23 Mpa
  • Tensile typical value: 32 Mpa
Offering superior comfort and exceptional grip,More Product Information