CellPro™ NZCYM Media, Molecular Biology Grade, Sterile Filtered
Supplier: Alkali Scientific
Prepare nutrient-rich media for bacterial growth with Alkali Scientific CellPro™ NZCYM Media, ensured sterile filtered for consistent results.
- Filter-sterilized, ready-to-use format
- Convenient package size
- Standard formulation
CellPro™ NZCYM Media is a nutrient-rich growth medium optimized for the cultivation of Escherichia coli strains. With its formulation of casein hydrolysate, yeast extract, and sodium chloride, this media supports rapid bacterial growth and high plasmid yield, making it ideal for molecular cloning and protein expression experiments. Through meticulous sterile filtration, CellPro™ NZCYM Media ensures purity and consistency, empowering researchers with reliable results in molecular biology workflows.