CellPro™ SOC Media, Molecular Biology Grade, Sterile Filtered
Supplier: Alkali Scientific
DC0011 DC0022 DC0002
104027-774EA 56.91 USD
104027-774 104028-620 104029-060
CellPro™ SOC Media, Molecular Biology Grade, Sterile Filtered
Cell Culture Media
Cultivate bacterial cultures with Alkali Scientific CellPro™ SOC Media, provided sterile filtered and meeting molecular biology grade standards.
  • For molecular biology
  • 0.2 μm filtered
  • Sterile
  • Ultra-pure
CellPro™ SOC Media is a specialized growth medium designed to support the transformation and propagation of Escherichia coli cells. Formulated with essential nutrients and salts, this media promotes rapid cell growth and high transformation efficiency, making it ideal for molecular cloning and recombinant DNA techniques. Through meticulous sterile filtration, CellPro™ SOC Media maintains purity and consistency, ensuring optimal performance in molecular biology laboratories.