PurePoint™ Rainin LTS™ LiteTouch™ Sterile Tips
Supplier: Alkali Scientific
SLT-1000 SLT-20 SLT-250
104029-070EA 53.83 USD
104029-070 104027-988 104028-584
PurePoint™ Rainin LTS™ LiteTouch™ Sterile Tips
Pipette Tips
Experience minimal sample retention with Rainin LTS™ LiteTouch™ sterile tips, designed for low retention and sterile applications.
  • Low forces - reduces tip ejection force up to 85%
  • Low retention - most precise, accurate delivery
  • Stable new rack designed for multichannel pipets
  • Great fit on Rainin®* LiteTouch™ pipets
  • RNAse, DNAse, pyrogen free
  • Sterile
The Rainin LTS™ LiteTouch™ sterile pipette tips - low retention are meticulously designed to cater to a wide range of pipetting needs, including compatibility with Rainin® LiteTouch™ pipets, LTS series, XTS series, and XLS series. These tips ensure the precise and accurate delivery of liquids with their low retention design, minimizing sample waste and enhancing experimental reproducibility. With an innovative rack designed for stability and compatibility with multichannel pipets, they offer seamless integration into various laboratory workflows.
Their reduced tip ejection force of up to 85% not only ensures ergonomic pipetting but also reduces the risk of repetitive strain injuries. Furthermore, being RNAse, DNAse, and Pyrogen-free, these sterile tips maintain the integrity of your samples, making them ideal for sensitive molecular biology applications.