PurePoint™ Reload Refillable Pipette Tips
Supplier: Alkali Scientific
RT1000 RT1010 RT1200 RT1250 RT1300
104029-044EA 43.06 USD
104029-044 104028-290 104028-100 104026-928 104028-044
PurePoint™ Reload Refillable Pipette Tips
Pipette Tips
Refill your pipette tip racks easily with Alkali Scientific™ PurePoint™ reload refillable pipette tips, ensuring accurate and reliable liquid handling in your experiments.
  • Earth friendly stackable wafer replacements - 80% less waste
  • Compatible with all PurePoint™ racks and your empty blue racks
  • Universal fit tips
  • Stackable wafer replacements
  • Convenient packaging
  • Reload tips match/pair with most empty tip boxes
  • Recommended autoclave procedure: 120 °C for 12 minutes at 15 psi
PurePoint™ reload pipette tip box are the economical choice when it comes toMore Product Information