Fetal Bovine Serum (FBS), USDA Approved
Supplier: Alkali Scientific
FB72 FB73
104028-456EA 544.6 USD
104028-456 104027-296
Fetal Bovine Serum (FBS), USDA Approved
Sera (Serum)
Maintain reliable cell culture conditions with Alkali Scientific™ Fetal Bovine Serum, USDA Approved, provided in a 500 ml volume.
  • Triple 0.1 µm sterile-filtered
  • Mycoplasma-tested and virus-screened
  • Ships on dry ice. Store frozen at −20 to −10 °C
CellPro™ Fetal Bovine Serum (FBS) approved by the United States Department of Agriculture (USDA) offers researchers a premium-grade serum product specifically optimized for cell culture applications in biomedical research. Sourced from USDA-approved abattoirs and processed under stringent quality controlMore Product Information