Fetal Bovine Serum (FBS), US Source
Supplier: Alkali Scientific
FB60 FB61
104026-678EA 956.91 USD
104026-678 104028-650
Fetal Bovine Serum (FBS), US Source
Sera (Serum)
Ensure consistent cell culture growth with Alkali Scientific™ Fetal Bovine Serum, sourced from US, available in a 500 ml volume.
  • Triple 0.1 µm sterile-filtered
  • Mycoplasma-tested and virus-screened
  • Ships on dry ice. Store frozen at −20 to −10 °C
CellPro™ Fetal Bovine Serum (FBS) sourced from the United States is a premium-quality serum product designed to support a wide range of cell culture applications in biomedical research. Derived from USDA-approved abattoirs and processed under stringent quality standards, this FBS product offers researchers aMore Product Information