PurePoint™ Rainin LTS™ LiteTouch™ Filter Tips
Supplier: Alkali Scientific
FLT-1000 FLT-20 FLT-200
104028-694EA 86.08 USD
104028-694 104027-020 104026-932
PurePoint™ Rainin LTS™ LiteTouch™ Filter Tips
Pipette Tips
Experience minimal sample loss with Alkali Scientific™ Rainin LTS™ LiteTouch™ filter tips - low retention, ensuring accurate pipetting.
  • Low forces - reduces tip ejection force up to 85%
  • Low retention - most precise, accurate delivery
  • Stable new rack designed for multichannel pipets
  • Great fit on Rainin®* LiteTouch™ pipets
  • RNAse, DNAse, pyrogen free
  • Sterile
Alkali Scientific™ presents PurePoint™ Rainin LTS™ LiteTouch™ filter tips - low retention, meticulously crafted for seamless compatibility with Rainin LTS™ and Rainin XLS™ pipettes, including multichannel pipettes. Engineered with precision and quality, these filter tips ensure optimal performance and compatibility with a wide range of Rainin pipettes, facilitating effortless sample handling and accurate dispensing.
The innovative low-retention design minimizes sample loss and maximizes recovery, making these tips ideal for various liquid handling applications in molecular biology, genomics, proteomics, and drug discovery. With their superior performance, versatility, and multichannel pipette friendliness, Alkali Scientific™ PurePoint™ Rainin LTS™ LiteTouch™ filter tips - low retention are the perfect choice for researchers seeking reliable and efficient sample processing in their laboratory workflows.