DEFENDER™ Safety Needle Holders, Covidien
Supplier: Covidien
DEFENDER™
8881225110
84009-736CS 569.07 USD
84009-736
DEFENDER™ Safety Needle Holders, Covidien
Blood Collection Tube Holders
These single-use devices eliminate needle removal and holder reuse, as well as provide front and back end needle protection.
  • They feature controlled needle retraction and do not require a technique change for ease of use
  • They also reduce sharps disposal cost by reducing the amount of space needed for disposal
Packaging: Packaged in bags of 20.