Manganese(II) sulfate monohydrate

Supplier: RPI
Danger

M85150-1000.0 M85150-500.0
77510-342EA 341.46 USD
77510-342 77510-344
Manganese(II) sulfate monohydrate
Manganese (II) sulphate monohydrate

Caution: Research or further manufacturing use only, not for food or drug use.

Formula: MnSO₄·1H₂O
Melting Pt: >449 °C
Density: 300…1200 kg/m³
Storage Temperature: Room Temperature
MDL Number: MFCD00149159
CAS Number: 10034-96-5
EINECS: 232-089-9

Order Now

Specification Test Results

Appearance Pale, pink fine granular powder
Identification (A) Meets the requirements
Identification (B) Meets the requirements
Assay 98.0 - 102.0%
Substances not precipitated by ammonium sulfide ≤0.5%
Loss on Ignition 10.0 - 13.0%
Grade USP
Production Method Chemical Synthesis
BSE/TSE Free BSE/TSE Free
Country of Origin IN


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR