Water, CellPro™, Tissue Culture Grade

Supplier: Alkali Scientific

Synonyms: Dihydrogen oxide

DTC01 DTC01-20L DTC01-4L
104027-638EA 41.52 USD
104027-638 104027-356 104027-700
Water, CellPro™, Tissue Culture Grade
Water

CellPro™ Tissue Culture Medical Grade Water stands as a trusted solution for cell culture applications. Crafted to meticulous standards of purity and sterility, this water ensures optimal conditions for cell growth and viability. With its sterile filtered formulation, CellPro™ Tissue Culture Medical Grade Water provides researchers with a reliable source of water for their experiments, facilitating successful outcomes in tissue culture studies.


  • For molecular biology
  • Liquid Medium
  • 0.2 μm filtered

Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
MDL Number: MFCD00011332
CAS Number: 7732-18-5
UN: 3248
Merck Index: 13,10098

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR