Tetracycline solution 15 mg/ml, CellPro™

Supplier: Alkali Scientific

DS0203 DS0186
104027-096EA 132.29 USD
104027-096 104026-892
Tetracycline solution 15 mg/ml, CellPro™
Tetracycline

CellPro™ Tetracycline Solution offers researchers a reliable antibiotic solution for molecular biology applications. Crafted to stringent standards of purity and sterility, this solution ensures consistent inhibition of bacterial growth in experimental procedures. With its sterile filtered formulation, CellPro™ Tetracycline Solution provides researchers with peace of mind and confidence in their work, facilitating successful outcomes in molecular biology studies.


  • For molecular biology
  • Liquid Medium
  • 0.2 μm filtered

Formula: C₂₂H₂₄N₂O₈
MDL Number: MFCD00151232
CAS Number: 60-54-8
UN: 3248

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR