D-(+)-Glucose solution 20%, CellPro™
Supplier: Alkali Scientific
CellPro™ Dextrose Solution stands as a cornerstone of excellence in molecular biology research. Crafted to meticulous standards of purity and sterility, this solution offers researchers a reliable source of glucose for cell culture applications. With its sterile filtered formulation, CellPro™ Dextrose Solution ensures consistent and reproducible results in experimental procedures, providing researchers with confidence and peace of mind in their work.
- For molecular biology
- Liquid Medium
- 0.2 μm filtered
Formula:
C₆H₁₂O₆ |
MDL Number:
MFCD00148912 CAS Number: 50-99-7 UN: 3248 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...