G418 disulfate solution 50 mg/ml, CellPro™

Supplier: Alkali Scientific

Synonyms: G-418 disulfateG-418 disulphateG418 disulfateG-418 sulphateG418 sulphateG-418 sulfateGeneticin®

DS0182
104028-018EA 147.68 USD
104028-018
G418 disulfate solution 50 mg/ml, CellPro™
G418 disulphate

CellPro™ G-418 Sulfate Solution represents the epitome of quality and reliability in molecular biology research. Crafted to stringent standards of purity and sterility, this solution offers researchers a potent antibiotic option for selection of transfected cells. With its sterile filtered formulation, CellPro™ G-418 Sulfate Solution ensures consistent and reproducible results in cell culture experiments, empowering researchers with confidence in their molecular biology endeavors.


  • For molecular biology
  • Liquid Medium
  • 0.2 μm filtered

Formula: C₂₀H₄₄N₄O₁₈S₂
MW: 692.72 g/mol
MDL Number: MFCD00058314
CAS Number: 108321-42-2
UN: 3248

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR