Chloramphenicol solution 10 mg/ml, CellPro™

Supplier: Alkali Scientific

Synonyms: Chloromycetin®(R,R)-ChloramphenicolTega-Cetin

DS0172
104027-032EA 87.68 USD
104027-032
Chloramphenicol solution 10 mg/ml, CellPro™
Chloramphenicol

CellPro™ Chloramphenicol Solution sets the standard for quality and reliability in molecular biology research. Formulated to stringent standards of purity and sterility, this solution provides researchers with a potent antibiotic option for selective bacterial culture. With a concentration of 10mg/mL in a 10mL volume, CellPro™ Chloramphenicol Solution ensures consistent and reproducible inhibition of bacterial growth, empowering researchers with confidence in their experimental procedures.


  • For molecular biology
  • Liquid Medium
  • 0.2 μm filtered

Formula: Cl₂CHCONHCH(CH₂OH)CH(OH)C₆H₄NO₂
MW: 323.13 g/mol
MDL Number: MFCD00078159
CAS Number: 56-75-7
UN: 3248

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR