Ampicillin solution 100 mg/ml, CellPro™
Supplier: Alkali Scientific
Synonyms:
D(-)-α-Aminobenzylpenicillin Sodium Salt, Ampicillin Na, Ampicillin sodium salt, D-(−)-α-Aminobenzylpenicillin sodium salt Ampicillin sodium salt
Alkali Scientific™ Ampicillin Solution offers researchers a high-quality antibiotic solution for molecular biology applications. Crafted to stringent standards of purity and sterility, this sterile filtered solution ensures reliable and consistent inhibition of bacterial growth in experimental procedures. Ideal for selective bacterial culture and plasmid maintenance, Alkali Scientific™ Ampicillin Solution guarantees optimal results and peace of mind in your laboratory work.
- For molecular biology
- Liquid Medium
- 0.2 μm filtered
Formula:
C₁₆H₁₈N₃NaO₄S MW: 371.39 g/mol |
MDL Number:
MFCD00064313 CAS Number: 69-52-3 UN: 3248 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...