Myricetin
Supplier: RPI
Synonyms:
3,3',4',5,5',7-Hexahydroxyflavone, 3,5,7-Trihydroxy-2-(3,4,5-trihydroxyphenyl)-4H-chromen-4-one
M95100-0.025
M95100-0.1
M95100-0.25
M95100-0.5
M95100-0.010
76347-152EA
136.63
USD
76347-152
76347-154
76347-156
76347-158
77510-346
Myricetin
Myricetin
Myricetin is a naturally occurring flavonol, a flavonoid found in many grapes, berries, fruits, vegetables, herbs as well as other plants.
Caution: Research or further manufacturing use only, not for food or drug use.
Formula:
C₁₅H₁₀O₈ MW: 318.24 g/mol Melting Pt: >300 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00006827 CAS Number: 529-44-2 EINECS: 208-463-2 |
Specification Test Results
Appearance | Light yellow powder |
Odor and taste | Characteristic |
Purity (HPLC) | ≥98.0% |
Identification (HNMR) | Consistent with the structure |
Sieve Analysis | 100% pass 80 mesh |
Loss on Drying | ≤2.0% |
Ash | ≤2.0% |
Total Heavy metals | <10 ppm |
Lead | ≤1.0 ppm |
Arsenic | ≤1.0 ppm |
Mercury (Hg) | ≤0.1 ppm |
Cadmium | ≤2.0 ppm |
BSE/TSE Free | BSE/TSE Free |
Country of Origin | Report |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...