Magnesium acetate tetrahydrate

Supplier: RPI

Synonyms: Acetic acid magnesium salt tetrahydrate, Magnesium di(acetate) tetrahydrate

M24100-1000.0 M24100-500.0 M24100-5000.0 M24100-10000.0
76346-884EA 100.65 USD
76346-884 76346-886 76346-888 77510-322
Magnesium acetate tetrahydrate
Magnesium acetate tetrahydrate

Caution: Research or further manufacturing use only, not for food or drug use.

Formula: (H₃CCOO)₂Mg·4H₂O
Melting Pt: 72...75 °C
Density: 1,454 g/cm³
Storage Temperature: Room Temperature
MDL Number: MFCD00149214
CAS Number: 16674-78-5
EINECS: 205-554-9

Order Now

Specification Test Results

Appearance White Crystals
Barium (Ba) ≤0.001%
Calcium (Ca) ≤0.01%
Chloride (Cl) ≤0.001%
DNase None Detected
Heavy Metals (as Pb) ≤0.0005%
Insoluble Matter ≤0.005%
Iron (Fe) ≤0.0005%
Manganese ≤0.001%
Potassium (K) ≤0.005%
Purity 98.0 - 102.0%
RNase None Detected
Sodium (Na) ≤0.005%
Strontium ≤0.005%
Sulfate ≤0.005%
Grade ACS/Molecular Biological Grade
Country of Origin Report


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR