Potassium Acetate
Supplier: RPI
P42000-2500.0 P42000-500.0 P42000-5000.0 P42000-10000.0
76347-816EA 222.18 USD
76347-816 76347-818 76347-820 77510-402
Potassium Acetate
Buffers Biological Buffers
In research laboratories, potassium acetate is utilized for various purposes. It is often employed in nucleic acid purification procedures, such as RNA extraction, where it helps precipitate nucleic acids from solution.
  • Solubility: Water
Potassium acetate is also used in protein purification techniques, such as ammonium sulfate precipitation, to selectively precipitate proteins from solution based on their solubility characteristics.
Furthermore, potassium acetate is utilized as a buffering agent in biochemical assays and cell culture media. Its buffering capacity helps maintain a stable pH environment, which is critical for many enzymatic reactions and cell culture processes.
Caution: Research or further manufacturing use only, not for food or drug use.