CryoLok Storage Vials, 2 ml, Polypropylene, Round bottom, Internal Sealing Ring, Graduated
Supplier: Alkali Scientific
C4611-B C4611-G C4611-R C4611-W C4611-Y
104027-354EA 116.91 USD
104027-354 104028-744 104027-858 104026-592 104027-464
CryoLok Storage Vials, 2 ml, Polypropylene, Round bottom, Internal Sealing Ring, Graduated
Vials
Store your precious samples securely with Alkali Scientific™ 2 ml CryoLok storage vials, featuring polypropylene construction, round bottom, internal sealing ring, and graduated markings.
  • Round bottom
  • Internal sealing ring
  • Graduated
  • Sterile
The 2 ml CryoLok storage vials are essential for reliable and secure sample storage in cryogenic conditions. Constructed from durable polypropylene, these vials feature a round bottom design with an internal sealing ring to prevent leakage and ensure sample integrity. With graduated markings for easy volume identification, these vials provide precise sample measurement and tracking. Suitable for long-term storage in ultra-low temperature environments, the CryoLok storage vials offer excellent resistance to extreme temperatures and chemical exposure. Their robust construction and sealing mechanism make them ideal for a wide range of cryopreservation applications, including cell culture, biobanking, and molecular biology research.