Ninhydrin

Supplier: RPI
Warning

Synonyms: 2,2-Dihydroxyindan-1,3-dionTrioxohydrindene monohydrateNinhydrin monohydrate2,2-Dihydroxy-1,3-indanedioneIndanetrione hydrate2,2-Dihydroxyindane-1,3-dione1,2,3-Indantrione monohydrate

N92000-10.0 N92000-1000.0 N92000-100.0
77510-370EA 1158.2 USD
77510-370 77510-368 77510-366
Ninhydrin
Ninhydrin

Caution: Research or further manufacturing use only, not for food or drug use.

Formula: C₉H₆O₄
MW: 178.14 g/mol
Melting Pt: 250 °C
Storage Temperature: Room Temperature
MDL Number: MFCD00003791
CAS Number: 485-47-2
EINECS: 207-618-1

Order Now

Specification Test Results

Appearance Pale yellow crystalline, free flowing powder
Assay ≥ 99.0 %
Identification (IR) Conforms
Suitability for detection of amino acids Passes test
pH (1 %) 4.6 - 5.6
Sulfated Ash ≤ 0.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR