Anti-IgG Goat Antibody (490)

Supplier: Advansta
R-05771-250
103254-832EA 113.08 USD
103254-832
Anti-IgG Goat Antibody (490)
Antibodies
Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: 490
Epitope:
Host: Goat
Isotype:
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR