Anti-IgG Goat Antibody (550)

Supplier: Advansta
R-05766-250
103254-814EA 113.08 USD
103254-814
Anti-IgG Goat Antibody (550)
Antibodies
Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: 490
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR