Arachidonic acid ≥99%

Supplier: Ambeed
Warning

Synonyms: (5Z,8Z,11Z,14Z)-Icosa-5,8,11,14-tetraenoic acid

A834351-1G A834351-5G A834351-100MG A834351-250MG
77552-554EA 185 USD
77552-554 77552-556 77573-658 77878-686
Arachidonic acid ≥99%
Arachidonic acid

Caution: for Research Use Only

Formula: C₂₀H₃₂O₂
MW: 304.5 g/mol
MDL Number: MFCD00004417
CAS Number: 506-32-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR