Ethoxy(pentafluoro)cyclotriphosphazene 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
2-Ethoxy-2,4,4,6,6-pentafluoro-1,3,5,2,4,6-triazatriphosphorine
A827856-10G
A827856-25G
A827856-5G
A827856-1G
77552-538EA
26
USD
77552-538
77552-540
77552-542
77569-544
Ethoxy(pentafluoro)cyclotriphosphazene 98%
Ethoxy(pentafluoro)cyclotriphosphazene
Caution: For research use only.
Formula:
C₂H₅F₅N₃OP₃ |
MDL Number:
MFCD28386107 CAS Number: 33027-66-6 UN: 1760 ADR: 8,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...