5-Benzylmercaptotetrazole (BMT, BTT) ≥98%

Supplier: Ambeed
Danger

Synonyms: BTT5-(Benzylthio)-1H-tetrazoleBMT

A791935-25G A791935-5G
77552-442EA 44.29 USD
77552-442 77552-444
5-Benzylmercaptotetrazole (BMT, BTT) ≥98%
5-(Benzylthio)-1H-tetrazole

Caution: for Research Use Only

Formula: C₈H₈N₄S
MW: 192.24 g/mol
MDL Number: MFCD00068731
CAS Number: 21871-47-6
UN: 1325
ADR: 4.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR