1-Vinyl-2-pyrrolidone ≥98% stabilized
Supplier: Ambeed
|
Danger
|
Synonyms:
N-Vinyl-2-pyrrolidinone
, N-Vinyl-2-pyrrolidone
Stabilization:
Stabilized with Ammonium hydroxide or NaOH 200 ppm
A790103-25G
77552-438EA
18
USD
77552-438
1-Vinyl-2-pyrrolidone ≥98% stabilized
1-Vinyl-2-pyrrolidone
Caution: for Research Use Only
Formula:
C₆H₉NO MW: 111.1418 g/mol |
MDL Number:
MFCD00003197 CAS Number: 88-12-0 UN: 2810 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...