N,N-Diethylhydroxylamine ≥98%

Supplier: Ambeed
Danger

A771380-25G A771380-100G
77552-382EA 23 USD
77552-382 77686-785
N,N-Diethylhydroxylamine ≥98%
N,N-Diethylhydroxylamine

Caution: for Research Use Only

Formula: C₄H₁₁NO
MDL Number: MFCD00002126
CAS Number: 3710-84-7
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR