tert-Butyl-4-methoxyphenol (mixture of isomers) 99+%

Supplier: Ambeed
Danger

Synonyms: tert-Butyl-4-methoxyphenol, BHA, tert-Butyl-4-hydroxyanisole, Butylhydroxyanisole, 2(3)-tert-Butyl-4-methoxyphenol, Butylated hydroxyanisole

A723359-25G A723359-10G
77552-238EA 8 USD
77552-238 77878-565
tert-Butyl-4-methoxyphenol (mixture of isomers) 99+%
tert-Butyl-4-methoxyphenol (mixture of isomers)

Caution: for Research Use Only

Formula: C₁₁H₁₆O₂
MW: 180.25 g/mol
Boiling Pt: 264
Flash Pt: 156
MDL Number: MFCD01779059
CAS Number: 25013-16-5
UN: 3077
ADR: 9 Ⅲ
Merck Index: 12,01582

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR