Bromocyclobutane ≥98%

Supplier: Ambeed
Danger

Synonyms: Cyclobutyl bromide

A719858-10G A719858-25G A719858-5G A719858-1G
77552-220EA 16 USD
77552-220 77552-222 77552-224 77602-850
Bromocyclobutane ≥98%
Bromocyclobutane

Caution: for Research Use Only

Formula: C₄H₇Br
MW: 135 g/mol
MDL Number: MFCD00001317
CAS Number: 4399-47-7
UN: 1993
ADR: 3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR