Methyl(phenyl)silane ≥95%

Supplier: Ambeed
Danger

A645491-5G A645491-25G
77551-958EA 127 USD
77551-958 77573-078
Methyl(phenyl)silane ≥95%
Methyl(phenyl)silane

Caution: for Research Use Only

Formula: C₇H₁₀Si
MW: 122.239 g/mol
MDL Number: MFCD00039787
CAS Number: 766-08-5
UN: 1993
ADR: 3 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR