4-Amino-2,6-dichloronicotinic acid ≥95%
Supplier: Ambeed
|
Warning
|
A601252-1G
A601252-100MG
A601252-250MG
77551-810EA
655
USD
77551-810
77673-618
77884-448
4-Amino-2,6-dichloronicotinic acid ≥95%
4-Amino-2,6-dichloronicotinic acid
Caution: for Research Use Only
Formula:
C₆H₄Cl₂N₂O₂ |
MDL Number:
MFCD19105212 CAS Number: 929288-22-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...