Tacrolimus ≥98%

Supplier: Ambeed
Danger

Synonyms: FR900506, Fujimycin, (3S,4R,5S,8R,9E,12S,14S,15R,16S,18R,19R,26aS)-5,6,8,11,12,13,14,15,16,17,18,19,24,25,26,26a-Hexadecahydro-5,19-dihydroxy-3-[(1E)-2-[(1R,3R,4R)-4-hydroxy-3-methoxycyclohexyl]-1-methylethenyl]-14,16-dimethoxy-4,10,12,18-tetramethyl-8-(2-propen-1-yl)-15,19-epoxy-3H-pyrido[2,1-c][1,4]oxaazacyclotricosine-1,7,20,21(4H,23H)-tetrone

A335856-5G A335856-1G A335856-1MG A335856-250MG
77550-962EA 863 USD
77550-962 77612-926 77685-960 77685-961
Tacrolimus ≥98%
Tacrolimus

Caution: for Research Use Only

Formula: C₄₄H₆₉NO₁₂
MDL Number: MFCD00869853
CAS Number: 104987-11-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR