Caprylaldehyde ≥99%

Supplier: Ambeed
Danger

Synonyms: Octanal, Octylaldehyde, Octaldehyde

A241023-25G A241023-100G
77550-554EA 12 USD
77550-554 77600-942
Caprylaldehyde ≥99%
Caprylaldehyde

Caution: for Research Use Only

Formula: C₈H₁₆O
MDL Number: MFCD00007029
CAS Number: 124-13-0
UN: 1191
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR