n-Dodecane ≥97%

Supplier: Ambeed
Danger

A224864-25G A224864-100G
77550-462EA 13 USD
77550-462 77626-138
n-Dodecane ≥97%
n-Dodecane

Caution: for Research Use Only

Formula: H₃C(CH₂)₁₀CH₃
MDL Number: MFCD00008969
CAS Number: 112-40-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR